Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIEZO1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP178537 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178537 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178537 Supplier Novus Biologicals Supplier No. NBP178537
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 16 publications

PIEZO1 Polyclonal specifically detects PIEZO1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PIEZO1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:25, Immunohistochemistry-Paraffin 1:400
Formulation PBS, 30% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q92508
Gene Alias FAM38A, member A, membrane protein induced by beta-amyloid treatment, Mib, piezo-type mechanosensitive ion channel component 1
Gene Symbols PIEZO1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human PIEZO1 protein (between residues 1300-1350) [UniProt Q92508]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9780
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.