Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIEZO2 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB337847 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337847 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337847 Supplier Novus Biologicals Supplier No. NBP178538APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PIEZO2 Polyclonal antibody specifically detects PIEZO2 in Human, Mouse, Porcine samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen PIEZO2
Applications Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias C18orf30, C18orf58, FAM38B, FAM38B2, family with sequence similarity 38, member B, family with sequence similarity 38, member B2, FLJ23144, FLJ23403, FLJ25916, FLJ34907, FLJ37734, FLJ45725, HsT748, HsT771, piezo-type mechanosensitive ion channel component 2, protein PIEZO2
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human PIEZO2 protein (between residues 1450-1500). [UniProt Q9H5I5]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 63895
Target Species Human, Mouse, Pig
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.