Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIEZO2 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NBP178538B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178538B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178538B Supplier Novus Biologicals Supplier No. NBP178538B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PIEZO2 Polyclonal specifically detects PIEZO2 in Human, Mouse, Porcine samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PIEZO2
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9H5I5
Gene Alias C18orf30, C18orf58, FAM38B, FAM38B2, family with sequence similarity 38, member B, family with sequence similarity 38, member B2, FLJ23144, FLJ23403, FLJ25916, FLJ34907, FLJ37734, FLJ45725, HsT748, HsT771, piezo-type mechanosensitive ion channel component 2, protein PIEZO2
Gene Symbols PIEZO2
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human PIEZO2 protein (between residues 1450-1500). [UniProt Q9H5I5]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 63895
Test Specificity Human and mouse. Porcine reactivity reported in scientific literature (PMID: 25385580).
Target Species Human, Mouse, Pig
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.