Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIASy Antibody, Novus Biologicals™
SDP

Catalog No. NB6001318SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001318SS 0.025 mg
NB6001318 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB6001318SS Supplier Novus Biologicals Supplier No. NB6001318SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 6 publications

PIASy Polyclonal specifically detects PIASy in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PIASy
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3 - 5 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:2000. Use reported in scientific literature (Ihara et al (2005))
Formulation PBS containing 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q8N2W9
Gene Alias E3 SUMO-protein ligase PIAS4, FLJ12419, MGC35296, Piasg, PIAS-gamma, PIASy, Protein inhibitor of activated STAT protein 4, Protein inhibitor of activated STAT protein gamma, protein inhibitor of activated STAT protein PIASy, protein inhibitor of activated STAT, 4, zinc finger, MIZ-type containing 6, ZMIZ6
Gene Symbols PIAS4
Host Species Rabbit
Immunogen This polyclonal antibody was developed against a mixture of synthetic peptides corresponding to amino acids 46-60 (KKIKELYETRYAKKNC) and 187-198 (YSDTSCPQEDQY) of human PIASg; GenBank no. AAD45155.1 AF164437_1. These sequences are 95% and 100% homologous in mouse and rat.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51588
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.