Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIF1 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP197931R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197931R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197931R Supplier Novus Biologicals Supplier No. NBP197931R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PIF1 Polyclonal specifically detects PIF1 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Western Blot (Negative), ICC/IF (Negative).

Specifications

Antigen PIF1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate DyLight 550
Dilution Immunohistochemistry 1:300, Immunohistochemistry-Paraffin 1:300, Western Blot (Negative), ICC/IF (Negative)
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q9H611
Gene Alias C15orf20, PIF, PIF1 5-to-3 Prime DNA helicase homolog, RRM3 DNA helicase-like protein
Gene Symbols PIF1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human PIF1 protein (between residues 400-500) [UniProt Q9H611]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 80119
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.