Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIF4 Rabbit anti-A. thaliana, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB167231 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167231 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB167231 Supplier Novus Biologicals Supplier No. NBP316068100UL
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PIF4 Polyclonal antibody specifically detects PIF4 in A. thaliana samples. It is validated for Western Blot

Specifications

Antigen PIF4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias At2g43010, AtbHLH9, AtPIF4, Basic helix-loop-helix protein 9, bHLH 9, bHLH transcription factor bHLH009, BHLH9, EN102, MFL8.13, Phytochrome-interacting factor 4, Short under red-light 2, SRL2, Transcription factor EN 102, Transcription factor PIF4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 300-400 of arabidopsis thaliana PIF4 (NP_565991.2).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 818903
Target Species A. thaliana
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.