Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIG3 Antibody (026), HRP, Novus Biologicals™
SDP

Catalog No. NB072115 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB072115 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB072115 Supplier Novus Biologicals Supplier No. NBP290309H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PIG3 Monoclonal specifically detects PIG3 in Human samples. It is validated for ELISA.

Specifications

Antigen PIG3
Applications ELISA
Classification Monoclonal
Clone 26
Conjugate HRP
Dilution ELISA
Formulation PBS
Gene Alias p53-induced gene 3 protein, PIG3quinone oxidoreductase homolog, quinone oxidoreductase PIG3, tumor protein p53 inducible protein 3, Tumor protein p53-inducible protein 3
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human PIG3 (NP_004872.2; Met1-Gln332).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 9540
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.