Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIG3 Antibody (OTI3B11), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP273400FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273400FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273400FR Supplier Novus Biologicals Supplier No. NBP273400FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PIG3 Monoclonal specifically detects PIG3 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen PIG3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3B11
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias p53-induced gene 3 protein, PIG3quinone oxidoreductase homolog, quinone oxidoreductase PIG3, tumor protein p53 inducible protein 3, Tumor protein p53-inducible protein 3
Gene Symbols TP53I3
Host Species Mouse
Immunogen Full length recombinant protein of human PIG3 (NP_671713) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 9540.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.