Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIGA Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159701
Description
PIGA Polyclonal specifically detects PIGA in Human samples. It is validated for Western Blot.Specifications
PIGA | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
class A GlcNAc-inositol phospholipid assembly protein, EC 2.4.1.198, GlcNAc-PI synthesis protein, GPI anchor biosynthesis, GPI3, phosphatidylinositol glycan anchor biosynthesis, class A, phosphatidylinositol glycan, class A (paroxysmal nocturnal hemoglobinuria), phosphatidylinositol N-acetylglucosaminyltransferase subunit A, Phosphatidylinositol-glycan biosynthesis class A protein, phosphatidylinositol-glycan biosynthesis, class A protein, PIG-A | |
Rabbit | |
19 kDa | |
100 μL | |
Primary | |
This product is specific to Subunit or Isoform: A. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Rabbit | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
B3KUV7 | |
PIGA | |
Synthetic peptides corresponding to PIGA(phosphatidylinositol glycan anchor biosynthesis, class A) The peptide sequence was selected from the middle region of PIGA. Peptide sequence SVKSLCEGLEKAIFQLKSGTLPAPENIHNIVKTFYTWRNVAERTEKVYDR. | |
Affinity purified | |
RUO | |
5277 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction