Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIGF Antibody, Novus Biologicals™
SDP

Catalog No. NBP169261 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP169261 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP169261 Supplier Novus Biologicals Supplier No. NBP169261
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PIGF Polyclonal specifically detects PIGF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PIGF
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q07326
Gene Alias GPI11 homolog, MGC32646, MGC33136, phosphatidylinositol glycan anchor biosynthesis, class F, phosphatidylinositol glycan, class F, phosphatidylinositol-glycan biosynthesis class F protein, PIG-F
Gene Symbols PIGF
Host Species Rabbit
Immunogen Synthetic peptides corresponding to PIGF(phosphatidylinositol glycan anchor biosynthesis, class F) The peptide sequence was selected from the N terminal of PIGF. Peptide sequence MKDNDIKRLLYTHLLCIFSIILSVFIPSLFLENFSILETHLTWLCICSGF.
Molecular Weight of Antigen 25 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5281
Test Specificity Porcine: 86%; Rat: 86%; Mouse: 86%; Sheep: 86%; Bovine: 86%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Sheep
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.