Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIGF Antibody (RM0010-8F09), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBT85468700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBT85468700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBT85468700 Supplier Novus Biologicals Supplier No. NB11085468AF700
Add to Cart
Add to Cart

Rat Monoclonal Antibody

PIGF Monoclonal specifically detects PIGF in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen PIGF
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone RM0010-8F09
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias GPI11 homolog, MGC32646, MGC33136, phosphatidylinositol glycan anchor biosynthesis, class F, phosphatidylinositol glycan, class F, phosphatidylinositol-glycan biosynthesis class F protein, PIG-F
Gene Symbols PIGF
Host Species Rat
Immunogen Mouse recombinant PlGF protein.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 5281
Test Specificity This recognizes mouse PlGF.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.