Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIGF Antibody (RM0010-8F09) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018036 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018036 0.025 mg
NB11085468 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018036 Supplier Novus Biologicals Supplier No. NB110854680.025MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

PIGF Monoclonal specifically detects PIGF in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen PIGF
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone RM0010-8F09
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:400-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:50-1:100
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Alias GPI11 homolog, MGC32646, MGC33136, phosphatidylinositol glycan anchor biosynthesis, class F, phosphatidylinositol glycan, class F, phosphatidylinositol-glycan biosynthesis class F protein, PIG-F
Gene Symbols PIGF
Host Species Rat
Immunogen Mouse recombinant PlGF protein.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5281
Test Specificity This recognizes mouse PlGF.
Reconstitution Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.