Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pIgR Antibody (40), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB333252 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333252 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333252 Supplier Novus Biologicals Supplier No. NBP290428APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

pIgR Monoclonal antibody specifically detects pIgR in Mouse samples. It is validated for ELISA, ELISA

Specifications

Antigen pIgR
Applications ELISA
Classification Monoclonal
Clone 40
Conjugate APC
Dilution ELISA, Sandwich ELISA Capture
Formulation PBS
Gene Alias FLJ22667, hepatocellular carcinoma associated protein TB6, Hepatocellular carcinoma-associated protein TB6, MGC125361, MGC125362, PIgR, Poly-Ig receptor, polymeric immunoglobulin receptor
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse pIgR (Accession#: NP_035212.2, Met 1-Lys 645)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 5284
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.