Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIGS Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen PIGS
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP174155
SDP
View Documents
Novus Biologicals
NBP174155
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

PIGS Polyclonal specifically detects PIGS in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

PIGS
Polyclonal
Rabbit
Q6PD26
94005
Synthetic peptides corresponding to thesis class S Antibody against the C terminal of Pigs. Immunizing peptide sequence AVAAVQKAAKALALGHLSSAFAASQEAVTSSERAFFDPSLLHLLYFPDDQ.
Primary
Western Blot
Unconjugated
RUO
DKFZp686K20216, FLJ45226, GPI transamidase component PIG-S, GPI transamidase subunit, phosphatidylinositol glycan anchor biosynthesis, class S, phosphatidylinositol glycan, class S, Phosphatidylinositol-glycan biosynthesis class S protein
PIGS
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.