Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIGW Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179309
Description
PIGW Polyclonal specifically detects PIGW in Rat samples. It is validated for Western Blot.Specifications
PIGW | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
class W, EC 2.3, FLJ37433, phosphatidylinositol glycan anchor biosynthesis, class W, PIG-W | |
Rabbit | |
Affinity purified | |
RUO | |
284098 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_919443 | |
PIGW | |
Synthetic peptide directed towards the middle region of human PigwThe immunogen for this antibody is Pigw. Peptide sequence SIGYQEHSTEYGVHWNFFFTIIVVKLITSLLLIIFPLNKSWIVAISITVL. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Rat: 92%; Bovine: 85%; Mouse: 85%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Yeast, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction