Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIH1D2 Antibody (OTI4A10), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N273403P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273403P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273403P594 Supplier Novus Biologicals Supplier No. NBP273403PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PIH1D2 Monoclonal specifically detects PIH1D2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen PIH1D2
Applications Flow Cytometry
Classification Monoclonal
Clone OTI4A10
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias PIH1 domain containing 2, PIH1 domain-containing protein 2
Gene Symbols PIH1D2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-315 of human PIH1D2 (NP_620144) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 120379.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.