Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIK3C2A Antibody (OTI3H2), PE, Novus Biologicals™
SDP

Catalog No. NB301961 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301961 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301961 Supplier Novus Biologicals Supplier No. NBP273405PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PIK3C2A Monoclonal antibody specifically detects PIK3C2A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen PIK3C2A
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3H2
Conjugate PE
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias C2-containing phosphatidylinositol kinase, CPK, DKFZp686L193, EC 2.7.1, EC 2.7.1.154, MGC142218, phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit alpha, Phosphoinositide 3-kinase-C2-alpha, phosphoinositide-3-kinase, class 2, alpha polypeptide, PI3-K-C2(ALPHA), PI3-K-C2A, PI3K-C2alpha, PI3K-C2-alpha, PtdIns-3-kinase C2 subunit alpha
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 230-560 of human PIK3C2A (NP_002636) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5286
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.