Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIK3C2G Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. p-7211228 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5124 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5124 Supplier Proteintech Group Inc Supplier No. 259041AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phosphoinositide 3-kinase activity is implicated in assorted cellular responses activated by mammalian cell surface receptors and the regulation of protein sorting in yeast. The p110gamma (PIK3CG) enzyme is activated in vitro by both the alpha and betagamma subunits of heterotrimeric GTP-binding proteins (G proteins) and does not associate with a p85 adaptor molecule. PI 3-kinase C2gamma, also designated p110gamma, may link signaling through G protein-coupled receptors to the generation of phosphoinositide second messengers that are phosphorylated in the D-3 position. The PI 3-kinase C2gamma gene encodes a 1,050 amino acid polypeptide with 36% identity to human PI 3-kinase C2alpha. Research indicates that PI 3-kinase C2gamma can block the growth of human colon cancer cells.

Specifications

Antigen PIK3C2G
Applications Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PIK3C2G
Gene Accession No. O75747
Gene Alias PI3K C2 gamma, PI3K C2GAMMA, PIK3C2G
Gene Symbols PIK3C2G
Host Species Rabbit
Immunogen PIK3C2G Fusion Protein Ag23177
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5288
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.