Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIK3R5 Antibody (OTI4G9), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP273408A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273408A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273408A70 Supplier Novus Biologicals Supplier No. NBP273408AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PIK3R5 Monoclonal specifically detects PIK3R5 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PIK3R5
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4G9
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias F730038I15Rik, FOAP-2, p101, P101-PI3K, Phosphatidylinositol-4,5-bisphosphate 3-kinase regulatory subunit, phosphoinositide-3-kinase, regulatory subunit 5, phosphoinositide-3-kinase, regulatory subunit, polypeptide p101, PI3-kinase p101 subunit, PI3-kinase regulatory subunit 5, PtdIns-3-kinase regulatory subunit, regulatory subunit 5, p101
Gene Symbols PIK3R5
Host Species Mouse
Immunogen Full length recombinant protein of human PIK3R5 (NP_055123) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 23533
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.