Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIK3R6 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP24856025UL

 View more versions of this product

Catalog No. NB396109


Only null left
Add to Cart

Description

Description

PIK3R6 Polyclonal antibody specifically detects PIK3R6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

PIK3R6
Polyclonal
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
C17orf38, chromosome 17 open reading frame 38, FLJ34500, HsT41028, p84, p87 phosphoinositide 3-kinase gamma (PI3Kg) adapter protein, p87 PI3K adapter protein, p87(PIKAP), p87PIKAPDKFZp666P158, Phosphoinositide 3-kinase gamma adapter protein of 87 kDa, phosphoinositide 3-kinase regulatory subunit 6, phosphoinositide-3-kinase, regulatory subunit 6, PI3Kgamma adapter protein of 87 kDa
This antibody was developed against a recombinant protein corresponding to amino acids: PAAPVTDHTCLNVNVTEVVKSSNLAGKSFSTVTNTFRTNNIQIQSRDQRLLTLSLDKDDQRTFRDVVRFEVAPCPEPCSGAQKSKAPWLNLHGQQEVEAIKAKPKP
25 μL
Protein Kinase, Signal Transduction
146850
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.2), 40% Glycerol
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.