Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIK3R6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | PIK3R6 |
---|---|
Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PIK3R6 Polyclonal antibody specifically detects PIK3R6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
PIK3R6 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Protein Kinase, Signal Transduction | |
PBS (pH 7.2), 40% Glycerol | |
146850 | |
IgG | |
Immunogen affinity purified |
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
C17orf38, chromosome 17 open reading frame 38, FLJ34500, HsT41028, p84, p87 phosphoinositide 3-kinase gamma (PI3Kg) adapter protein, p87 PI3K adapter protein, p87(PIKAP), p87PIKAPDKFZp666P158, Phosphoinositide 3-kinase gamma adapter protein of 87 kDa, phosphoinositide 3-kinase regulatory subunit 6, phosphoinositide-3-kinase, regulatory subunit 6, PI3Kgamma adapter protein of 87 kDa | |
This antibody was developed against a recombinant protein corresponding to amino acids: PAAPVTDHTCLNVNVTEVVKSSNLAGKSFSTVTNTFRTNNIQIQSRDQRLLTLSLDKDDQRTFRDVVRFEVAPCPEPCSGAQKSKAPWLNLHGQQEVEAIKAKPKP | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title