Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tingley™ Pilot™ General Purpose PVC Knee Boots

Ideal for use in construction, agriculture, general purpose and foul weather applications. Tingley™ Pilot™ General Purpose PVC Knee Boots provide easy walking as well as all day comfort.

Specifications

Certifications/Compliance Prop 65
Color Black
Design Pull-on
Footbed Material PVC
For Use With (Application) Construction, agriculture, and light industrial
View More Specs

Products 10
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-161-331
View Documents
Tingley™
3115114
14 N/A N/A N/A
19-161-328
View Documents
Tingley™
3115113
13 N/A N/A N/A
19-161-326
View Documents
Tingley™
3115112
12 N/A N/A N/A
19-161-324
View Documents
Tingley™
3115111
11 N/A N/A N/A
19-161-323
View Documents
Tingley™
3115110
10 N/A N/A N/A
19-161-321
View Documents
Tingley™
3115109
9 N/A N/A N/A
19-161-319
View Documents
Tingley™
3115108
8 N/A N/A N/A
19-161-318
View Documents
Tingley™
3115107
7 N/A N/A N/A
19-161-317
View Documents
Tingley™
3115106
6 N/A N/A N/A
19-161-316
View Documents
Tingley™
3115105
5 N/A N/A N/A
Description

Description

  • Plain Toe
  • Injection molded construction for 100% waterproof protection
  • Flexible upper material stays supple in cold temperatures to make walking easier
  • Tougher outsole material for good abrasion resistance
  • Self-cleaning cleated outsole spits out debris as you walk
  • Removable contour EVA insole wicks perspiration to help keep feet dry and comfortable
Specifications

Specifications

Prop 65
Pull-on
Construction, agriculture, and light industrial
Injection Molded
PVC
Cleated
6
Yes
Black
PVC
EVA Insole
Self Cleaning
Knee Boot
Reusable
Chemicals
Boot
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.