Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PILR-alpha Antibody (2175B), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB346980 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346980 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346980 Supplier Novus Biologicals Supplier No. FAB64842MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PILR-alpha Monoclonal antibody specifically detects PILR-alpha in Human samples. It is validated for Flow Cytometry, Neutralization

Specifications

Antigen PILR-alpha
Applications Flow Cytometry, Neutralization
Classification Monoclonal
Clone 2175B
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, Neutralization
Formulation 50mM Sodium Borate
Gene Alias Cell surface receptor FDF03, FDF03, inhibitory receptor PILRalpha, Inhibitory receptor PILR-alpha, paired immunoglobin-like receptor alpha, paired immunoglobin-like type 2 receptor alpha, paired immunoglobulin-like receptor alpha, paired immunoglobulin-like type 2 receptor alpha
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived recombinant human PILR-alpha with a C-terminal 6-His tag, Gln20-Thr196, Accession # Q9UKJ1
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 29992
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.