Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PILR-alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP16009520UL
Description
PILR-alpha Polyclonal specifically detects PILR-alpha in Human samples. It is validated for Western Blot.Specifications
PILR-alpha | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
Q9UKJ1-4 | |
PILRA | |
Synthetic peptides corresponding to PILRA(paired immunoglobin-like type 2 receptor alpha) The peptide sequence was selected from the N terminal of PILRA. Peptide sequence IPFSFYYPWELATAPDVRISWRRGHFHRQSFYSTRPPSIHKDYVNRLFLN. | |
20 μL | |
Signal Transduction | |
29992 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
Cell surface receptor FDF03, FDF03, inhibitory receptor PILRalpha, Inhibitory receptor PILR-alpha, paired immunoglobin-like receptor alpha, paired immunoglobin-like type 2 receptor alpha, paired immunoglobulin-like receptor alpha, paired immunoglobulin-like type 2 receptor alpha | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction