Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PILR-alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | PILR-alpha |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP16009520
![]() |
Novus Biologicals
NBP16009520UL |
20 μL |
Each for $206.00
|
|
|||||
NBP160095
![]() |
Novus Biologicals
NBP160095 |
100 μL |
Each for $487.50
|
|
|||||
Description
PILR-alpha Polyclonal specifically detects PILR-alpha in Human samples. It is validated for Western Blot.Specifications
PILR-alpha | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
Q9UKJ1-4 | |
29992 | |
Synthetic peptides corresponding to PILRA(paired immunoglobin-like type 2 receptor alpha) The peptide sequence was selected from the N terminal of PILRA. Peptide sequence IPFSFYYPWELATAPDVRISWRRGHFHRQSFYSTRPPSIHKDYVNRLFLN. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
Cell surface receptor FDF03, FDF03, inhibitory receptor PILRalpha, Inhibitory receptor PILR-alpha, paired immunoglobin-like receptor alpha, paired immunoglobin-like type 2 receptor alpha, paired immunoglobulin-like receptor alpha, paired immunoglobulin-like type 2 receptor alpha | |
PILRA | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title