Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIM2 Antibody (OTI5D5), PerCP, Novus Biologicals™
SDP

Catalog No. NB273412PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB273412PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB273412PCP Supplier Novus Biologicals Supplier No. NBP273412PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PIM2 Monoclonal specifically detects PIM2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen PIM2
Applications Flow Cytometry
Classification Monoclonal
Clone OTI5D5
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 2.7.11.1, pim-2 oncogene, pim-2h, proto-oncogene Pim-2 (serine threonine kinase), serine/threonine protein kinase pim-2, serine/threonine-protein kinase pim-2
Gene Symbols PIM2
Host Species Mouse
Immunogen Full length recombinant protein of human PIM2 (NP_006866) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 11040.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.