Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIM2 Rabbit anti-Human, Clone: 3V1S5, Novus Biologicals™
SDP

Catalog No. NB168670 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168670 20 μg
NB168669 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB168670 Supplier Novus Biologicals Supplier No. NBP31678920UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PIM2 Monoclonal antibody specifically detects PIM2 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen PIM2
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 3V1S5
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EC 2.7.11.1, pim-2 oncogene, pim-2h, proto-oncogene Pim-2 (serine threonine kinase), serine/threonine protein kinase pim-2, serine/threonine-protein kinase pim-2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 10-120 of human PIM2 (Q9P1W9).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 11040
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.