Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIM2 Rabbit anti-Human, Clone: 3V1S5, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | PIM2 |
---|---|
Clone | 3V1S5 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
PIM2 Monoclonal antibody specifically detects PIM2 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
PIM2 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Protein Kinase | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
11040 | |
IgG | |
Affinity purified |
3V1S5 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 2.7.11.1, pim-2 oncogene, pim-2h, proto-oncogene Pim-2 (serine threonine kinase), serine/threonine protein kinase pim-2, serine/threonine-protein kinase pim-2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 10-120 of human PIM2 (Q9P1W9). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title