Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIMT Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen PIMT
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP154942
SDP
View Documents
Novus Biologicals
NBP154942
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

PIMT Polyclonal specifically detects PIMT in Human samples. It is validated for Western Blot.
Specifications

Specifications

PIMT
Polyclonal
Rabbit
Transcription Factors and Regulators
Cap-specific guanine-N2 methyltransferase, CLL-associated antigen KW-2, DKFZp762A163, EC 2.1.1, EC 2.1.1.-, HCA137, Hepatocellular carcinoma-associated antigen 137, NCOA6IPSEREX-defined, nuclear receptor coactivator 6 interacting protein, Nuclear receptor coactivator 6-interacting protein, PIMTFLJ22995, PIPMT, PRIP-interacting protein PIPMT, PRIP-interacting protein with methyltransferase domain, PRIP-interacting protein with methyltransferase motif, trimethylguanosine synthase, trimethylguanosine synthase 1, trimethylguanosine synthase homolog, trimethylguanosine synthase homolog (S. cerevisiae)
TGS1
IgG
96 kDa
Western Blot
Unconjugated
RUO
Q96RS0
96764
Synthetic peptides corresponding to TGS1(trimethylguanosine synthase homolog (S. cerevisiae)) The peptide sequence was selected from the middle region of TGS1. Peptide sequence IDENPASDFDDSGSLLGFKYGSGQKYGGIPNFSHRQVRYLEKNVKLKSKY.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.