Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIN/DLC8 Rabbit anti-Human, Clone: SD08-04, Novus Biologicals™
SDP

Catalog No. NBP267796 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267796 Supplier Novus Biologicals Supplier No. NBP267796
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PIN/DLC8 Monoclonal specifically detects PIN/DLC8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen PIN/DLC8
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SD08-04
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias cytoplasmic dynein light polypeptide, DLC1DNCLC1, DLC8MGC126137, DNCL1MGC126138, dynein light chain 1, cytoplasmic, Dynein light chain LC8-type 1, dynein, cytoplasmic, light polypeptide 1, dynein, light chain, LC8-type 1,8 kDa dynein light chain, hdlc1, LC8, PINLC8a, Protein inhibitor of neuronal nitric oxide synthase
Gene Symbols DYNLL1
Host Species Rabbit
Immunogen Synthetic peptide within Human PIN/DLC8 aa 1-50/ 89. (SwissProt: P63167 Human; SwissProt: P63168 Mouse; SwissProt: P63170 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline p53 Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8655
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.