Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pin1 Antibody (3G8) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP122956 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP122956 0.1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP122956 Supplier Novus Biologicals Supplier No. NBP122956
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pin1 Monoclonal specifically detects Pin1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, SDS-Page.

Specifications

Antigen Pin1
Applications Western Blot, Flow Cytometry, ELISA, SDS-Page
Classification Monoclonal
Clone 3G8
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Gene Accession No. NP_006212
Gene Alias dod, EC 5.2.1.8, peptidyl-prolyl cis/trans isomerase, NIMA-interacting, peptidylprolyl cis/trans isomerase, NIMA-interacting 1, peptidyl-prolyl cis-trans isomerase NIMA-interacting 1, Peptidyl-prolyl cis-trans isomerase Pin1, PPIase Pin1, prolyl isomerase, protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1, protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1, Rotamase Pin1, UBL5
Gene Symbols PIN1
Host Species Mouse
Immunogen Recombinant human Pin1 (1-163 aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 ml
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Mitotic Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5300
Test Specificity Anti-human Pin1 mAb, clone 3G8, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Pin1 protein.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.