Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pin1 Antibody, Novus Biologicals™

Chicken Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP1054240.025ML
Description
Pin1 Polyclonal specifically detects Pin1 in Human, Mouse, Rat, Porcine, Bovine, Chicken, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Pin1 | |
Polyclonal | |
Western Blot 1:10000, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunohistochemistry-Paraffin | |
dod, EC 5.2.1.8, peptidyl-prolyl cis/trans isomerase, NIMA-interacting, peptidylprolyl cis/trans isomerase, NIMA-interacting 1, peptidyl-prolyl cis-trans isomerase NIMA-interacting 1, Peptidyl-prolyl cis-trans isomerase Pin1, PPIase Pin1, prolyl isomerase, protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1, protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1, Rotamase Pin1, UBL5 | |
Chicken | |
21 kDa | |
0.025 mL | |
Cell Cycle and Replication, Mitotic Regulators | |
5300 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgY |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry, Western Blot, Immunofluorescence | |
Unconjugated | |
PBS with 10mM Sodium Azide | |
PIN1 | |
Recombinant full length Pin1 purified from E. coli. | |
Ammonium sulfate precipitation | |
RUO | |
Primary | |
Pig, Rat, Bovine, Mouse, Chicken, Equine, Human | |
Precipitation |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction