Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pin1 Antibody, Novus Biologicals™

Chicken Polyclonal Antibody has been used in 2 publications
$206.50 - $499.50
Specifications
Antigen | Pin1 |
---|---|
Dilution | Western Blot 1:10000, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunohistochemistry-Paraffin |
Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry, Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Pin1 Polyclonal specifically detects Pin1 in Human, Mouse, Rat, Porcine, Bovine, Chicken, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Pin1 | |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry, Western Blot, Immunofluorescence | |
Unconjugated | |
Chicken | |
Cell Cycle and Replication, Mitotic Regulators | |
PBS with 10mM Sodium Azide | |
5300 | |
Recombinant full length Pin1 purified from E. coli. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 1:10000, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunohistochemistry-Paraffin | |
Polyclonal | |
Precipitation | |
RUO | |
Pig, Rat, Bovine, Mouse, Chicken, Equine, Human | |
dod, EC 5.2.1.8, peptidyl-prolyl cis/trans isomerase, NIMA-interacting, peptidylprolyl cis/trans isomerase, NIMA-interacting 1, peptidyl-prolyl cis-trans isomerase NIMA-interacting 1, Peptidyl-prolyl cis-trans isomerase Pin1, PPIase Pin1, prolyl isomerase, protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1, protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1, Rotamase Pin1, UBL5 | |
PIN1 | |
IgY | |
Ammonium sulfate precipitation | |
21 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title