Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Pinacidil Monohydrate K+ channel activator
Shop All R&D Systems Products

Click to view available options
Quantity:
10 mg
50 mg
Description
PubChem ID: 55329
Application Notes: May be dissolved in DMSO (25 mg/mL, warm); or ethanol (25 mg/mL, warm)
Specifications
Specifications
| CAS | 85371-64-8 |
| Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Molecular Formula | C13H19N5· H2O |
| Purity | >98% (HPLC); NMR (conforms) |
| Quantity | 10 mg |
| Source | Synthetic |
| Target | Pinacidil Monohydrate K+ channel activator |
| Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction