Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pinin Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB291881 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291881 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291881 Supplier Novus Biologicals Supplier No. NB10055315APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Pinin Polyclonal antibody specifically detects Pinin in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen Pinin
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias 140 kDa nuclear and cell adhesion-related phosphoprotein, Desmosome-associated protein, Domain-rich serine protein, DRS protein, DRSP, DRSSR-like protein, Melanoma metastasis clone A protein, MEMA, neutrophil protein, Nuclear protein SDK3, pinin, pinin, desmosome associated protein, SDK3
Host Species Rabbit
Immunogen Maps to a region between residue 667 and the C-terminus (residue 717) of human Pinin, desmosome associated protein using the numbering given in entry NP_002678.2 (GeneID 5411).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5411
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.