Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PINK1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB355226 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355226 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355226 Supplier Novus Biologicals Supplier No. BC100494APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PINK1 Polyclonal specifically detects PINK1 in Human, Mouse, Rabbit, Rat samples. It is validated for Electron Microscopy, Gene Knock-Out, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Peptide ELISA, SDS-Page, Western Blot.

Specifications

Antigen PINK1
Applications Electron Microscopy, Gene Knock-Out, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Peptide ELISA, SDS-Page, Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Peptide ELISA, Electron Microscopy, SDS-Page, Knockout Validated, Knockdown Validated
Formulation PBS
Gene Alias BRPK, EC 2.7.11.1, FLJ27236, PARK6, Parkinson disease (autosomal recessive) 6, protein kinase BRPK, PTEN Induced Kinase 1, PTEN induced putative kinase 1, PTEN-induced putative kinase protein 1, serine/threonine-protein kinase PINK1, mitochondrial
Host Species Rabbit
Immunogen PINK1 antibody was developed using a synthetic peptide made to the human PINK1 protein sequence (between residues 175-250). [Swiss-Prot Q9BXM7]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Autophagy, Cardiovascular Biology, DNA Repair, Lipid and Metabolism, Mitochondrial Markers, Mitophagy, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 65018
Target Species Human, Mouse, Rabbit, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.