Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega PinPoint™ Vector Sequencing Primer
Shop All Promega Corporation Products
Click to view available options
Quantity:
2 μg
Description
- Hybridizes upstream of the Factor Xa site at nucleotides 325–343, approximately 40–50 base pairs upstream of the multiple cloning region
- Can be used to determine if an insert is cloned in-frame with the biotinylation purification tag of the PinPoint Xa Vectors
- Sequence: 5'-d(CGTGACGCGGTGCAGGGCG)-3'
- Tested in double-stranded sequencing reactions with circular PinPoint Vectors
- Lyophilized
- Antibody Search Quantity: 2μg
- Size: 2μg
Specifications
Specifications
| Content And Storage | Store at −20°C. |
| Description | PinPoint |
| Product Type | Primer |
| Quantity | 2 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction