Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PINX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | PINX1 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7931
![]() ![]() |
Proteintech Group Inc
123681AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Valosin containing protein (VCP), also designated TERA (for transitional endoplasmic reticulum ATPase), is a member of the AAA family of ATPases, which are involved in a variety of cellular activities. VCP is involved in a variety of membrane functions and in the regulation of the cell cycle. VCP associates with ubiquitinated IkB-a as well as with the 26S proteosome, indicating a potential role for VCP in the proteosome-mediated degradation of IkB-a. NVL (nuclear valosin-containing protein-like), also known as NVLp, is an 856 amino acid nuclear protein belonging to the AAA ATPase family. Implicated in ATP-dependent nuclear processes and ribosome synthesis, NVL exists as three alternatively spliced isoforms designated NVL isoform 1 (NVLp.2), NVL isoform 2 (NVLp.1) and NVL isoform 3. Widely expressed, NVL is found at highest levels in pancreas, retina, heart, skeletal muscle and placenta.Specifications
PINX1 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
A4L691, Q96BK5, Q9CZX5 | |
305963, 54984, 72400 | |
PINX1 Fusion Protein Ag3043 | |
Primary | |
-20°C | |
PINX1 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
LPTL, LPTS, PIN2 interacting protein 1, Pin2 interacting protein X1, PINX1, Protein 67 11 3, TRF1 interacting protein 1 | |
PINX1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title