Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP Antibody (PIP/1571), Novus Biologicals™
SDP

Catalog No. NB25322620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB25322620 20 μg
NB253226100 100 μg
NB253226200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB25322620 Supplier Novus Biologicals Supplier No. NBP25322620UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PIP Monoclonal specifically detects PIP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.

Specifications

Antigen PIP
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page
Classification Monoclonal
Clone PIP/1571
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page
Formulation 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Gene Alias GCDFP15prolactin-inducible protein, GCDFP-15SABP, GPIP4gp17, Gross cystic disease fluid protein 15, prolactin-induced proteinSecretory actin-binding protein
Gene Symbols PIP
Host Species Mouse
Immunogen Recombinant human GCDFP-15 protein fragment (aa 41-146) (exact sequence is proprietary)
Molecular Weight of Antigen 15 kDa
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5304
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.