Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ ArmorLite™ Textured Nitrile Coated Cotton Gloves
Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Medium
Large
X-Large
Description
- Cotton interlock liner thinly coated with blue nitrile
- Comfortable and flexible for a high degree of dexterity
- Textured coating provides enhanced grip capabilites
- Effective, affordable protection from physical hazards such as cut, puncture, snags and abrasions
- Resistant to grease and oils
- Made in ISO 9001:2000 / ISO 14001:2000 facilities
- Launderable
Specifications
Specifications
| Coated Area | Palm, Fingers, Knuckles |
| Coating Material | Nitrile |
| Color | Blue, Natural |
| Exterior Finish | Partially Textured |
| Hand | Specific |
| Latex Free | Yes |
| Material | Cotton |
| Product Type | Dip-coated Gloves |
| Size | X-Large |
| For Use With (Application) | General Handling, Assembly, Fabrication, Inspection, and Maintenance |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction