Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ MaxiCut™ Dyneema™ Cut-Resistant Gloves, Nitrile Foam Coating

Catalog No. 19165656C Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Length (English):
8.1 in.
8.7 in.
9.1 in.
9.4 in.
Length (Metric):
20.5 cm
22 cm
23 cm
24 cm
Size:
Small
Medium
Large
X-Large
Glove Cuff Color:
Orange
Red
White
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Length (Metric) Size Glove Cuff Color
19-165-656C 9.1 in. 23 cm Large White
19-165-656A 8.1 in. 20.5 cm Small Red
19-165-656B 8.7 in. 22 cm Medium Orange
19-165-656D 9.4 in. 24 cm X-Large Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-165-656C Supplier PIP™ Supplier No. 19D470/L
This item is not returnable. View return policy

Seamless knit cut-resistant gloves with micro-foam nitrile coating are compatible with light oils and will provide a secure grip and abrasion resistance.

Ideal for use in electronics, glass cutting, metal handling, food processing and meat processing — resistant to chemicals, water and ultraviolet light

  • Micro-foam nitrile coatings are compatible with light oils and provide secure grip, abrasion resistance and overall breathability
  • Reinforcement between thumb and forefinger improves cut resistance and glove life
  • Knit wrists prevent dirt and debris from entering glove
  • Material resists chemicals and UV light
  • Seamless nylon shell incorporates glass, Dyneema™ and Lycra™
  • Micro-cup finish allows for a controlled grip
  • “Hand at rest” form reduces fatigue
  • Color-coded cuff
  • Washable

Certifications

The Oeko-Tex© Association has evaluated and certified this product as skin safe from the second it comes into contact with the skin.

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) CE
Construction Seamless Knit
Color Gray
Cut Resistance Rating ANSI A5
Material Nylon, Glass, Dyneema™, Lycra
Product Type Cut-resistant Gloves
Size Large
EN388 Standard EN 388:4443C
For Use With (Application) Electronics, glass cutting, metal handling, food processing and meat processing
Glove Cuff Color White
Glove Cuff Style Continuous Knit
Hem Color White
Model 19-D470/L
Puncture Rating ANSI/ISEA 105-16 Level 3
Series MaxiCut™ 19-D470
Yarn Gauge 13
Length (English) 9.1 in.
Length (Metric) 23 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.