Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Cabaret™ Cotton Dress Gloves, Dotted Palm with Raised Stitching - Open Cuff

Catalog No. 191500723 Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Constructed in full fashion pattern with fourchettes and set-in thumbs for a tailored fit. Commonly used for formal occasions by police, firemen, waiters, marching bands, military, and pallbearers.

  • Three lines of raised stitching on back
  • Open cuff allows for easy on and off
  • Dotted palm for enhanced grip capabilities

Specifications

Construction String Knit
Accelerator Free Yes
Applications Industrial
Coating Material Plastic Dots
Color White
Disposable Reusable
Exterior Finish Grip Pattern
Hand Specific
Material Cotton
Powdered Powder Free
Product Type Dress Gloves
Size X-Large
Sterility Non-sterile
Glove Cuff Color White
Glove Cuff Style Open
Palm Type Dotted
Series 130-100WMPD
Thumb Style Keystone
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.