Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ CleanTeam™ Seamless Knit Nylon/Carbon Fiber ESD Gloves

Catalog Number 19121206B
Click to view available options
Size:
Small
Medium
This item is not returnable. View return policy
This item is not returnable. View return policy

Electrostatic Dissipative (ESD) fiber yarns blended with low lint nylon reduce static build-up on the glove's surface for improved performance in electronics assembly.

  • Electrostatic Dissipative (ESD) fiber yarns blended with low lint nylon
  • Reduced static build-up on the glove's surface for improved performance in electronics assembly
  • Electrical Surface Resistivity (ASTM D 257:2007): Below 7.5 x 107
  • Seamless knit construction enhances dexterity
  • Launderable without losing ESD properties
  • Reusable
Construction Seamless Knit
Coating Material Uncoated
Color Gray
Disposable Reusable
Material Nylon, Carbon Fiber
Product Type ESD Gloves
Size Medium
For Use With (Application) Aerospace, Circuit Board Manufacturing, Electronics Assembly, and Munitions
Glove Cuff Style Continuous Knit
Hem Color Orange
Series 40-6410
Yarn Gauge 15
Length (English) 7.7 in.
Length (Metric) 19.5 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit