Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Superior Grade Top Grain Cowhide Leather Drivers Gloves - Gauntlet Cuff

Catalog No. 19053681 Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Large
X-Large
This item is not returnable. View return policy

Catalog No. 19-053-681

Supplier: PIP™ 68101G/L

This item is not returnable. View return policy

Top grain cowhide leather provides comfort, durability, abrasion resistance, and breathability. The plasticized gauntlet cuff extends protection past the wrist and allows for quick removal.

  • Cowhide leather provides comfort, durability, abrasion resistance, and breathability
  • Top grain leather is smooth, provides durability and dexterity
  • Elastic closure sewn inside glove for safety and comfort
  • Drivers gloves feature slip-on styling to provide good fit and comfort
  • Straight thumb increases comfort in closed-fisted work
  • Color coded hems for easy size identification
  • For use in construction, machine operation, light duty welding, utility work, farming, roping, and many other general applications

Specifications

Color Natural
Grade Superior
Hand Specific
Material Top Grain Cowhide Leather
Product Type Driver Gloves
Size Large
For Use With (Application) Construction, Machine Operation, Light Duty Welding, Utility Work, Farming, Roping, and Many Other General Applications
Glove Cuff Style Gauntlet
Hem Color Brown
Lining Unlined
Series 88-101G
Thumb Style Straight
Length (English) 9.8 in.
Length (Metric) 24.8 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.