Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Regular Grade Top Grain Cowhide Leather Drivers Gloves with Pull Strap

Catalog No. 19069486 Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Top grain cowhide leather provides comfort, durability, abrasion resistance, and breathability. Leather pull strap closure with metal buckle for a secure, personal fit.

  • Top grain leather is smooth, provides durability and dexterity
  • Drivers gloves feature slip-on styling to provide good fit and comfort
  • Straight thumb increases comfort in closed-fisted work
  • Meets the content requirements for Chromium VI Free Leather
  • Color coded hems for easy size identification
  • For use in construction, machine operation, light duty welding, utility work, farming, roping and many other general applications

Specifications

Color Natural
Grade Regular
Hand Specific
Material Top Grain Cowhide Leather
Product Type Driver Gloves
Size Large
Style Standard
For Use With (Application) Construction, Machine Operation, Light Duty Welding, Utility Work, Farming, Roping, and Many Other General Applications
Glove Cuff Style Open
Hem Color Brown
Lining Unlined
Series 68-153
Thumb Style Straight
Length (English) 9.7 in.
Length (Metric) 24.8 cm
Weight 39 lb. (Case)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.