Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Excalibur™ Nitrile Coated Cotton Driver's Gloves

Catalog No. p-3348749 Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Small
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Ideal for handling metal or wood sheets or objects and general maintenance.

  • Nitrile-coated cotton laminated fabric provides great snag, cut and abrasion resistance
  • Slip-on cuff allows for easy on and off
  • Gunn cut pattern delivers excellent durability, wear and comfort
  • Wing thumb increases comfort in open-handed work
  • Cotton lining for superior comfort
  • Fully laminated palm with fabric back keeps hands cool

  • Automotive assembly operations
  • Handling metal or wood sheets
  • Objects with sharp edges
  • General plant maintenance

Specifications

Back Material Fabric
Coating Material Nitrile
Color Brown, Neutral
Hand Specific
Product Type Dip-coated Gloves
Size X-Large
For Use With (Application) Automotive Assembly, Metal/Wood Sheet Handling, Sharp Objects, and General Plant Maintenance
Glove Cuff Style Open
Hem Color Blue
Liner Material Cotton
Series 60-3140
Length (English) 9.9 in.
Length (Metric) 25 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.