Learn More
PIP™ G-Tek™ Aramid Fiber Cut-Resistant Gloves, Nitrile Microfinish Grip
Shop All Protective Industrial Products Inc. ProductsDescription
- Longer wrist length for added protection
- Heavier and thicker shell provides enhanced protection against cuts
- Long-lasting and durable for great economic efficiency
- Double dipped MicroFinish™ coating provides remarkable comfort to the wearer and prevents wet hands, keeping the inside of the glove dry
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 4 |
| Construction | Seamless Knit |
| Coating Material | Nitrile |
| Color | Yellow, Black |
| Cut Resistance Rating | ANSI A3 |
| Material | Aramid Fiber |
| Product Type | Cut-resistant Gloves |
| Size | Medium |
| EN388 Standard | EN 388:4342B |
| For Use With (Application) | Material handling, glass handling operations, fishing industries, sanitation, general maintenance, woodworking, waste handling and recycling |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.