Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ CR Ultra Foam Nitrile Coated Gloves with Dyneema™ Fibers

Catalog No. p-4077803 Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy

Catalog No. 19-013-604

Supplier: PIP™ 19D855/L

This item is not returnable. View return policy

Dyneema™ fibers are 10 to 15 times stronger than steel, and more durable than polyester with a strength that is 40% greater than aramid fiber. Foam nitrile coating is compatible with light oils for enhanced grip capabilities.

  • Foam nitrile coated palm and knuckles are compatible with light oils and provide a good grip and excellent abrasion resistance
  • Seamless knit construction offers increased comfort and breathability
  • Dyneema™/nylon blend shell provides dexterity, tactile sensitivity, and cut resistance
  • Knit Wrist helps prevent dirt and debris from entering the glove
  • Resistant to chemicals, water and ultraviolet light

Specifications

Coated Area Palm, Fingers, Knuckles
Coating Material Nitrile Foam
Color Black, Gray
Cut Resistance Rating ANSI A2
Exterior Finish No Texture
Hand Specific
Material Dyneema, Nylon
Product Type Dip-coated Gloves
Size Large
For Use With (Application) Glass Handling, Injection Molding, Sheet Metal Operations, and Automotive Manufacturing
Glove Cuff Style Continuous Knit
Series 19-D855
Length (English) 9.8 in.
Length (Metric) 25 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.