Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ GP™ Premium Grade Crinkle Latex Coated Cotton/Polyester Gloves

Catalog No. 19151290 Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Latex crinkle coatings feature a surface texture that is designed to channel fluids away from the glove, allowing for better contact on wet and dry surfaces.

  • Seamless cotton/polyester knit shell is comfortable and flexible
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Ergonomic shaped curved finger pattern
  • Color coded hems for easy size identification
  • Ideal for handling/cleanup of abrasive parts and materials, demolition, masonry, metal fastening, and assembly

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Latex
Color Natural, Blue
Exterior Finish Fully Textured
Grade Premium
Hand Specific
Material Polycotton
Product Type Seamless Knit Gloves
Size 2X-Large
For Use With (Application) Handling/Cleanup of Abrasive Materials, Demolition, Masonry, Tile Work, and Metal Fastening
Glove Cuff Style Knit
Series 39-C1300
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.