Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ MaxiFlex™ Endurance™ Seamless Knit Nylon Gloves with Full Nitrile MicroFoam Grip

Catalog No. p-4893596 Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Raised micro dots increase durability and offer additional cushioning in highly repetitive applications that require precision handling. MicroFoam Nitrile coating is compatible with light oils for enhanced grip and abrasion resistance.

  • MicroFoam Nitrile coating is compatible with light oil to enhance grip and abrasion resistance
  • Raised micro dots increase durability and offer additional cushioning in highly repetitive applications that require precision handling
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Patented micro-foam nitrile coating offers 360° breathability, making it the most breathable glove on the market today
  • 25% thinner than most foam nitrile gloves while still offering twice the mechanical performance
  • Glove design mimics the hand at rest to reduce hand fatigue and increase comfort
  • Producted using the latest knitting technology for smooth and rounded fingertips, improving fingertip sensitivity
  • Consistent liner penetration from the nitrile leaves only the soft liner resting against the skin
  • Micro-cup finish allows for a controlled grip in all types of environments
  • All ingredients used in the manufacturing and construction of this product are compliant with the REACH legislation
  • Gloves are laundered prior to final packaging to ensure they are clean and safe for use
  • The Oeko-Tex™ Association has evaluated and certified this product as skin safe from the second it comes into contact with the skin
  • The Skin Health Alliance has awarded professional dermatological accreditation following review of the scientific dossier behind this product
  • 100% silicone free

Specifications

ANSI Abrasion Rating ANSI 3
Coated Area Full Glove
Construction Seamless Knit
Coating Material Nitrile
Color Black
Exterior Finish Fully Textured
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Medium
EN388 Standard 4131
For Use With (Application) General Assembly, Micro-Engineering, Automotive Components, Painting, and Horticulture
Glove Cuff Color Gray
Glove Cuff Style Continuous Knit
Palm Type Raised Micro-Dots
Series 34-846
Yarn Gauge 15
Length (English) 8.9 in.
Length (Metric) 22.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.